H-Phe-2-Cltrt Resin 1 * 5 g
Catalog # 04-12-2809-5
Supplier: Novabiochem (Part of Merck)
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Centrifuge tubes, Falcon®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Serological pipettes, Stripette™
Supplier: Corning
Petri dishes, Corning® Gosselin™
Supplier: Corning
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
Inoculating loops and needles, disposable
Supplier: Greiner Bio-One
Cryoboxes, 133×133 mm
Supplier: TENAK
Eppendorf Tubes® Microtubes with Screw Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection