Fmoc-Thi-OH
1 g
Catalog # 04-12-1082-1
Supplier: Novabiochem (Part of Merck)
Jump to:
Product Details & Documents
Synonyms: Fmoc-3-(2-thienyl)-L-alanine, Fmoc-β-(2-thienyl)-Ala-OH, (S)-2-(Fmoc-amino)-3-(2-thienyl)propionic acid, N-α-Fmoc-β-thienyl-L-alanine
- CAS Number:130309-35-2
- Formula:C₂₂H₁₉NO₄S
- MDL Number:MFCD00065676
- Molecular Weight:393.47 g/mol
Specifications
Specifications
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



