1-Bromopentane 98%
Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:110-53-2
- Formula:BrH₂C(CH₂)₃CH₃
- MDL Number:MFCD00000267
- Molecular Weight:151.05 g/mol
- Merck Index:13,00606
- Boiling Pt:129 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:1,21 g/cm³
- Flash Pt:79 °C
- Storage Temperature:Ambient
- EINECS:203-776-0
- UN:1993
- ADR:3,III
Documents
ACRO15252
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4430 to 1.4460 (20°C, 589 nm)
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:250 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



