5-Hydroxy-L-tryptophan 99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
5-Hydroxy-L-tryptophan 99%
Catalog # ACRO148290050
Supplier: Thermo Fisher Scientific
CAS Number: 4350-09-8
Jump to:
Product Details & Documents
Synonyms: (S)-2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid, 5-Hydroxyl-L-tryptophan, (-)-5-Hydroxy-L-Tryptophane, L-5-Hydroxytryptophan, Oxitriptan, H-Trp(5-OH)-OH
- CAS Number:4350-09-8
- Formula:C₁₁H₁₂N₂O₃
- MDL Number:MFCD00064341
- Molecular Weight:220.23 g/mol
- Merck Index:12,04895
- Melting Pt:270 °C (dec.)
- Storage Temperature:Ambient
- EINECS:224-411-1
- UN:2811
- ADR:6.1,III
Documents
ACRO14829
SDS
Specifications
Specification Test Results
- Appearance:White to light beige to light grey Powder
- Infrared spectrum:Conforms
- Water:≤1 % (K.F.)
- Heavy metals:≤10 ppm
- Sulfated ash:≤0.3 %
- HPLC:≤0.2 % (on dry substance) L-trypthophan
- Specific optical rotation:-30.9° to -34.9° (20°C, 589 nm) (c=1, H2O) (on dry substance)
- Titration with HClO4:98.5 to 101.5 % (On dry substance)
- Arsenic (As):≤1 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



