Cell standard rectangular with lid matched 10mm pathlength 10mm ID 2ml working volume 12,5x45mm external dimensions 1 * 8 items
80-2109-80
: Cytiva
Cell standard rectangular with lid matched 10mm pathlength 10mm ID 2ml working volume 12,5x45mm external dimensions 1 * 8 items
80-2109-80
: Cytiva
Frequently Bought Together
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection