Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
1,5-Pentanediol 98%

1,5-Pentanediol 98%

Glass bottle for solids, 100 g
Catalog # ACRO129951000
1,5-Pentanediol 98%
Catalog # ACRO129951000
CAS Number:  111-29-5
1,5-Pentanediol 98%
1,5-Pentanediol 98%

Glass bottle for solids, 100 g

CAS Number:  111-29-5
Price
£35.10
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Pentane-1,5-diol, 1,5-Dihydroxy pentane
  • CAS Number:111-29-5
  • Formula:C₅H₁₂O₂
  • MDL Number:MFCD00002978
  • Molecular Weight:104.15 g/mol
  • Merck Index:12,07256
  • Boiling Pt:242 °C (760 mmHg)
  • Melting Pt:–18 °C
  • Density:0,994 g/cm³
  • Flash Pt:135 °C
  • Storage Temperature:Ambient
  • EINECS:203-854-4

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4490 to 1.4510 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Elemental Analysis:
    51.20 to 55.47 % C
  • Residue after evaporation:
    ≤0.01 %
  • Peroxides:
    ≤50 ppm

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications