1,5-Pentanediol 98%
Glass bottle for solids, 1 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Pentane-1,5-diol, 1,5-Dihydroxy pentane
- CAS Number:111-29-5
- Formula:C₅H₁₂O₂
- MDL Number:MFCD00002978
- Molecular Weight:104.15 g/mol
- Merck Index:12,07256
- Boiling Pt:242 °C (760 mmHg)
- Melting Pt:–18 °C
- Density:0,994 g/cm³
- Flash Pt:135 °C
- Storage Temperature:Ambient
- EINECS:203-854-4
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4490 to 1.4510 (20°C, 589 nm)
- GC:≥97.5 %
- Elemental Analysis:51.20 to 55.47 % C
- Residue after evaporation:≤0.01 %
- Peroxides:≤50 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



