1,8-Octanediol 98%
Plastic bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,8-Dihydroxy octane
- CAS Number:629-41-4
- Formula:C₈H₁₈O₂
- MDL Number:MFCD00002989
- Molecular Weight:146.23 g/mol
- Boiling Pt:278,8 °C (760 mmHg)
- Melting Pt:60…62 °C
- Density:0,941 g/cm³ (20 °C)
- Flash Pt:128 °C
- Storage Temperature:Ambient
- EINECS:211-090-8
Documents
ACRO12938
SDS
Specifications
Specification Test Results
- Appearance:Colorless to white Powder or crystals
- Infrared spectrum:Conforms
- Melting point:58 to 63 °C
- Water:≤0.5 % (K.F.)
- GC:≥97.5 %
Specifications
- Pack type:Plastic bottle
- Pk:25 g



