3'-Nitroacetophenone 98%
Plastic bottle, 500 g
: m-Nitroacetophenone
- :121-89-1
- :C₈H₇NO₃
- :MFCD00007259
- :165.15 g/mol
- :202 °C (1013 hPa)
- :76…79 °C
- :1,433 g/cm³ (20 °C)
- :Ambient
- :204-504-3
Documents
ACRO12833
SDS
- Appearance:Yellow to yellow-green to yellow-brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:76 to 80 °C
- GC:≥97.5 %
- Pack type:Plastic bottle
- Pk:500 g



