3'-Nitroacetophenone 98%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
3'-Nitroacetophenone 98%
Catalog # ACRO128331000
Supplier: Thermo Fisher Scientific
CAS Number: 121-89-1
Jump to:
Product Details & Documents
Synonyms: m-Nitroacetophenone
- CAS Number:121-89-1
- Formula:C₈H₇NO₃
- MDL Number:MFCD00007259
- Molecular Weight:165.15 g/mol
- Boiling Pt:202 °C (1013 hPa)
- Melting Pt:76…79 °C
- Density:1,433 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-504-3
Documents
ACRO12833
SDS
Specifications
Specification Test Results
- Appearance:Yellow to yellow-green to yellow-brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:76 to 80 °C
- GC:≥97.5 %
Specifications
- Pack type:Plastic bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



