2'-Nitroacetophenone 95%
Glass bottle for solids, 20 g
: o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone
- :577-59-3
- :C₈H₇NO₃
- :MFCD00007144
- :165.15 g/mol
- :159 °C (16 mmHg)
- :28…30 °C
- :1,24 g/cm³ (20 °C)
- :>112 °C
- :Refrigerator
- :209-414-8
- Appearance:Orange to brown Low melting solid
- Infrared spectrum:Conforms
- GC:≥94 %
- Stabilizer:0.05 % BHT
- Pack type:Glass bottle for solids
- Pk:20 g



