2'-Nitroacetophenone 95%
Glass bottle for solids, 2,5 g
Supplier: Thermo Fisher Scientific
2'-Nitroacetophenone 95%
Catalog # ACRO128320025
Supplier: Thermo Fisher Scientific
CAS Number: 577-59-3
Jump to:
Product Details & Documents
Synonyms: o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone
- CAS Number:577-59-3
- Formula:C₈H₇NO₃
- MDL Number:MFCD00007144
- Molecular Weight:165.15 g/mol
- Boiling Pt:159 °C (16 mmHg)
- Melting Pt:28…30 °C
- Density:1,24 g/cm³ (20 °C)
- Flash Pt:>112 °C
- Storage Temperature:Refrigerator
- EINECS:209-414-8
Documents
ACRO12832
SDS
Specifications
Specification Test Results
- Appearance:Orange to brown Low melting solid
- Infrared spectrum:Conforms
- GC:≥94 %
- Stabilizer:0.05 % BHT
Specifications
- Pack type:Glass bottle for solids
- Pk:2,5 g



