N-Methylaniline 99%
Glass bottle for solids, 1 kg
- :100-61-8
- :C₇H₉N
- :MFCD00008283
- :107.16 g/mol
- :12,06098
- :195 °C (760 mmHg)
- :–57 °C
- :0,987 g/cm³
- :86 °C
- :Ambient
- :202-870-9
- :2294
- :6.1,III
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5685 to 1.5725 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤20 APHA
- Residue after evaporation:≤0.003 %
- Carbonyl compounds:≤0.01 % (as formaldehyde)
- Titrable acid:≤0.001 meq/g
- Pack type:Glass bottle for solids
- Pk:1 kg



