Mowiol 4-88 1 * 100 g
Catalog # 475904-100
Supplier: Merck Millipore (Calbiochem)
Frequently Bought Together
1-Chlorobutane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Supplier: Honeywell Chemicals
1,4-Dioxane 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Sodium hexanitritocobaltate(III)
Supplier: Thermo Fisher Scientific
Mineral oil, white stabilised USP, J.T.Baker®
Supplier: Avantor
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Alcohol, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide 99.9+% ACS
Supplier: Thermo Fisher Scientific
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific