Ethyl pyruvate 98%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester
- CAS Number:617-35-6
- Formula:C₅H₈O₃
- MDL Number:MFCD00009123
- Molecular Weight:116.12 g/mol
- Boiling Pt:144…147 °C (1013 hPa)
- Melting Pt:–58 °C
- Density:1,0453 g/cm³
- Flash Pt:45 °C
- Storage Temperature:Refrigerator
- EINECS:210-511-2
- UN:3272
- ADR:3,III
Documents
ACRO11894
SDS
Specifications
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4060 (20°C, 589 nm)
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



