2,3-Dimethoxybenzaldehyde 97%
Glass bottle for solids, 50 g
- :86-51-1
- :C₉H₁₀O₃
- :MFCD00003309
- :166.18 g/mol
- :137 °C (12 mmHg)
- :49…54 °C
- :>110 °C
- :Ambient
- :201-677-7
Documents
ACRO11537
SDS
- Appearance:Off-white to beige Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:48 to 53 °C
- GC:≥96.0 %
- Pack type:Glass bottle for solids
- Pk:50 g



