ContactUsLinkComponent
Alcoholometer, EUROPA classII, measuring range:30-40:0.1%vol, without thermometer, length:360mm, reference temperature: 20°C, suitab le for government verification. 1 * 1 items
Catalog # AMARH848116
Supplier:  Amarell
Alcoholometer, EUROPA classII, measuring range:30-40:0.1%vol, without thermometer, length:360mm, reference temperature: 20°C, suitab le for government verification. 1 * 1 items
Catalog # AMARH848116
Supplier:  Amarell
Frequently Bought Together