ContactUsLinkComponent
Alcoholometer, EUROPA classII, measuring range:0-10:0.1%vol, without thermometer, length:360mm, reference temperature: 20°C, suitabl e for government verification. 1 * 1 items
  AMARH848110
 :  Amarell
Alcoholometer, EUROPA classII, measuring range:0-10:0.1%vol, without thermometer, length:360mm, reference temperature: 20°C, suitabl e for government verification. 1 * 1 items
  AMARH848110
 :  Amarell
Frequently Bought Together