ContactUsLinkComponent
Areometer, DIN12791, L50 series, measuring range: 1.65-1.70:0.0005g/cm , without thermometer, reference temperature:20C, length:335 mm, suitable for government verification. 1 * 1 items
Catalog # AMARH806242
Supplier:  Amarell
Areometer, DIN12791, L50 series, measuring range: 1.65-1.70:0.0005g/cm , without thermometer, reference temperature:20C, length:335 mm, suitable for government verification. 1 * 1 items
Catalog # AMARH806242
Supplier:  Amarell
Frequently Bought Together