ContactUsLinkComponent
Areometer, DIN12791, L50 series, measuring range: 1.45-1.50:0.0005g/cm , without thermometer, reference temperature:20C, length:335 mm, suitable for government verification. 1 * 1 items
Catalog # AMARH806234
Supplier:  Amarell
Areometer, DIN12791, L50 series, measuring range: 1.45-1.50:0.0005g/cm , without thermometer, reference temperature:20C, length:335 mm, suitable for government verification. 1 * 1 items
Catalog # AMARH806234
Supplier:  Amarell
Frequently Bought Together