Areometer, DIN12791, L50 series, measuring range: 1.35-1.40:0.0005g/cm , without thermometer, reference temperature:20C, length:335 mm, suitable for government verification. 1 * 1 items
Catalog # AMARH806230
Supplier: Amarell
Frequently Bought Together