Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Flatpack chambers

Flatpack chambers

Flat pack chamber, 1,83 mm gap, 1,5 ml, 50
Catalog # 700-4813
Supplier:   BTX
Flatpack chambers
Catalog # 700-4813
Supplier:  BTX
CAS Number:  
Flatpack chambers
Flatpack chambers

Flat pack chamber, 1,83 mm gap, 1,5 ml, 50

CAS Number:  
Price
£636.00
Log in for availability
1 * 50 items
Product Details & Documents

Flatpack chambers are primarily used for prokaryotic applications (bacterial or yeast transformations), however they are also often used for high efficiency stem cell transfection.

  • The flow-through construction of the 0,56 mm gap has a volume capacity from 10 to 85 μl. This design provides the unique combination of small sample volumes with field strengths as high as 40 kV/cm.

The flatpack chamber with 1,83 mm gap has a three-ply solid sandwich construction of stainless steel and mylar plastic and holds a volume of 1,5 ml, ideal for stems cells.

Flatpack chambers are gamma sterilised in individual packages.

 : Supplied in sets of 50 and may be used in the BTX safety stand (available separately, please enquire).

Specifications

Specifications

  • Pk:
    50
  • Description:
    Flat pack chamber, 1,83 mm gap, 1,5 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.