Orders LinkContactUsLinkComponent
Magnetic stirring bars, PTFE, octagonal, with centre ring
Magnetic stirring bars, PTFE, octagonal, with centre ring
15 mm, 8 mm
Catalog # BRND137608
Supplier:   Brand
Magnetic stirring bars, PTFE, octagonal, with centre ring
Catalog # BRND137608
Supplier:  Brand
Magnetic stirring bars, PTFE, octagonal, with centre ring
Magnetic stirring bars, PTFE, octagonal, with centre ring
15 mm, 8 mm
Price
£53.30
Log in for availability
1 * 10 items
Product Details & Documents
These octagonal shaped PTFE magnetic stirrer bars are ideal for vessels with slightly uneven bottoms. The angular surface creates high turbulence even at low speeds.
Designed with pivot ring structure which reduces the contact surface and enables the stirrer to adopt the optimum position.
Specifications
  • Ø:
    8 mm
  • Length:
    15 mm
  • Seq:
    02