(R)-(-)-Epichlorhydrin 99%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
(R)-(-)-Epichlorhydrin 99%
Catalog # ACRO349010010
Supplier: Thermo Fisher Scientific
CAS Number: 51594-55-9
Jump to:
Product Details & Documents
Synonyms: (R)-(-)-Glycerol epichlorhydrin, (R)-(-)-3-Chloro-1,2-epoxypropane, (R)-(-)-1-Chloro-2,3-epoxypropane
- CAS Number:51594-55-9
- Formula:ClCH₂CH(O)CH₂
- MDL Number:MFCD00077759
- Molecular Weight:92.52 g/mol
- Boiling Pt:114 °C (760 mmHg)
- Melting Pt:–48 °C
- Density:1,183 g/cm³
- Flash Pt:33 °C
- Storage Temperature:Ambient
- EINECS:424-280-2
- UN:2023
- ADR:6.1,II
Documents
ACRO34901
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4360 to 1.4400 (20°C, 589 nm)
- GC:≥98.5 %
- Specific optical rotation:-34° to -39° (20°C, 589 nm) (c=1, MeOH)
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



