β-Cyclodextrine hydrate 99%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
β-Cyclodextrine hydrate 99%
Catalog # ACRO227281000
Supplier: Thermo Fisher Scientific
CAS Number: 68168-23-0
Jump to:
Product Details & Documents
- CAS Number:68168-23-0
- Formula:C₄₂H₇₀O₃₅·H₂O
- MDL Number:MFCD00150811
- Molecular Weight:1134.98 g/mol (anhy)
- Melting Pt:298…300 °C
- Storage Temperature:Ambient
Documents
ACRO22728
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤14 % (K.F.)
- Heavy metals:≤10 ppm
- Sulfated ash:≤0.1 %
- Solubility:(1 % in water) Clear colorless
- HPLC:≥98.5 % (ELSD)
- Related substances:≤1.0 % Alpha- and gamma-cyclodextrin (HPLC)
- pH:5.0 to 8.0 (1 % in water)
- Specific optical rotation:+155° to +167° (25°C, 589 nm) (c=1, H20) (On dry substance)
- Reducing substances:≤0.2 % (as Glucose)
- Residual solvents:≤5 ppm Toluene
Specifications
- Pack type:Glass bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



