β-Cyclodextrine hydrate 99%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
β-Cyclodextrine hydrate 99%
Catalog # ACRO227280250
Supplier: Thermo Fisher Scientific
CAS Number: 68168-23-0
Jump to:
Product Details & Documents
- CAS Number:68168-23-0
- Formula:C₄₂H₇₀O₃₅·H₂O
- MDL Number:MFCD00150811
- Molecular Weight:1134.98 g/mol (anhy)
- Melting Pt:298…300 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤14 % (K.F.)
- Heavy metals:≤10 ppm
- Sulfated ash:≤0.1 %
- Solubility:(1 % in water) Clear colorless
- HPLC:≥98.5 % (ELSD)
- Related substances:≤1.0 % Alpha- and gamma-cyclodextrin (HPLC)
- pH:5.0 to 8.0 (1 % in water)
- Specific optical rotation:+155° to +167° (25°C, 589 nm) (c=1, H20) (On dry substance)
- Reducing substances:≤0.2 % (as Glucose)
- Residual solvents:≤5 ppm Toluene
Specifications
- Pack type:Glass bottle
- Pk:25 g



