Volumetric flasks, class A, white markings
10 ml, 7/16, 0,025 ± ml
Catalog # 612-3467
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, amber, with glass stopper, conformity certified (DE-M marking).
- White enamelled indelible markings resistant to acids and alkalis
- Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck.
Certifications: DIN EN ISO 1042.
Documents
Specifications
- NS:7/16
- Capacity:10 ml
- Tolerance:0,025 ± ml