ContactUsLinkComponent
Reader,Rotoreader,for 24 micro-haematocrit tubes while on centrifuge 1 * 1 items
Catalog # HAWK01560-00
Supplier:  HAWKSLEY AND SONS
Reader,Rotoreader,for 24 micro-haematocrit tubes while on centrifuge 1 * 1 items
Catalog # HAWK01560-00
Supplier:  HAWKSLEY AND SONS
Frequently Bought Together