Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
1,1-Diphenylethane 96%

1,1-Diphenylethane 96%

Bottle
Catalog # APOSOR1027572-250M
New Product
Supplier:   Apollo Scientific
1,1-Diphenylethane 96%
Catalog # APOSOR1027572-250M
Supplier:  Apollo Scientific
New Product
CAS Number:  612-00-0
1,1-Diphenylethane 96%

Bottle

CAS Number:  612-00-0
Price
£49.80
Log in for availability
1 * 250 mg
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:612-00-0
  • Formula:C₁₄H₁₄
  • Molecular Weight:182,26 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.