16-Mercaptohexadecanoic acid 98%
Bottle
Catalog # APOSOR1027258-1G
New Product
Supplier: Apollo Scientific
16-Mercaptohexadecanoic acid 98%
Catalog # APOSOR1027258-1G
Supplier: Apollo Scientific
New Product
CAS Number: 69839-68-5
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: 16-Mercaptohexadecanoic acid
- CAS Number:69839-68-5
- Formula:C₁₆H₃₂O₂S
- Molecular Weight:288,49 g/mol
Specifications
Specifications
- Pack type:Bottle
- Cat. No.:APOSOR1027258-1G
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



