Orders LinkContactUsLinkComponent
3,5-Dimethylcyclohexanone 95%
Bottle
  APOSOR938239-100G
New Product
3,5-Dimethylcyclohexanone 95%
  APOSOR938239-100G
New Product
 :  2320-30-1
3,5-Dimethylcyclohexanone 95%
Bottle
 :  2320-30-1
£800.00
1 * 100 g
  •  :2320-30-1
  •  :C₈H₁₄O
  •  :MFCD00019467
  •  :126.1962 g/mol
  •  :Ambient
  • Pack type:
    Bottle