ContactUsLinkComponent
Screw cap, made of ABS, black, for 50ml ointment jar with thread 51mm 1 * 25 items
Catalog # RIXI8-1090-51
Supplier:  RIXIUS
New Product
Screw cap, made of ABS, black, for 50ml ointment jar with thread 51mm 1 * 25 items
Catalog # RIXI8-1090-51
Supplier:  RIXIUS