Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

Dutasteride ≥99% dual 5alpha reductase type 1 and type 2 inhibitor

Marketsource
Supplier:   Abcam
Dutasteride ≥99% dual 5alpha reductase type 1 and type 2 inhibitor
Supplier:  Abcam
Marketsource
Certificates
Dutasteride ≥99% dual 5alpha reductase type 1 and type 2 inhibitor

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Selective, competitive dual 5α reductase type 1 and type 2 inhibitor. Blocks the conversion of testosterone into dihydrotestosterone. Shows antitumor effects in vivo. Orally active.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Number:164656-23-9
  • Molecular Weight:528,5 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% (5a,17ß)-N-[2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-aza-androst-1-ene-17-carboxamide
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - £56.40
    £56.40
    1 * 10 mg
       £56.40

    Specifications:
    Marketsource
    Marketsource
  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - £231.00
    £231.00
    1 * 50 mg
       £231.00

    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.