Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Cell Lysis Buffer, Abcam

Cell Lysis Buffer, Abcam

Supplier:   Abcam
Cell Lysis Buffer, Abcam
Supplier:  Abcam
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

This product is manufactured by BioVision, an Abcam company and was previously called 1067 Cell Lysis Buffer.

  • Constituents: 1% Alcohol Ethoxylate, 0.88% Sodium chloride, 0.32% Tris HCl

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ABCAAB288311-100
  • ABCAAB288311-400
  • Storage temperature:
  • Store at +4°C.
  • Store at +4°C.

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • ABCAAB288311-100
    Cell Lysis Buffer
    Cell Lysis Buffer
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - £108.00
    £108.00
    1 * 100 mL
       £108.00

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB288311-100
    Storage temperature
    Store at +4°C.
  • ABCAAB288311-400
    Cell Lysis Buffer
    Cell Lysis Buffer
    Log in for availability
    Log in for availability
    1 * 400 mL
    1 * 400 mL - £221.00
    £221.00
    1 * 400 mL
       £221.00

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB288311-400
    Storage temperature
    Store at +4°C.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.