ContactUsLinkComponent
G-Biosciences Tris-EDTA-NaCl Buffer
G-Biosciences Tris-EDTA-NaCl Buffer
Catalog # GENO786-901
Supplier:  G-Biosciences
CAS Number:  
G-Biosciences Tris-EDTA-NaCl Buffer
Catalog # GENO786-901
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Description:
    Tris-EDTA-NaCl buffer, pH 8,0 (10 mM Tris, 1 mM EDTA, 50 mM NaCl)
  • Pk Info:
    1 L
Specifications
About this item
 Tris-EDTA-NaCl buffer solution at pH 8,0.
  • 10 mM Tris, 1 mM EDTA, 50 mM NaCl
  • Precision in concentration and pH
  • Guaranteed reproducibility
A versatile, mild buffer system widely used in molecular biology for DNA/RNA stabilisation, extraction, and storage.