ContactUsLinkComponent
Vent Assembly, 2 Port, Straight
Vent Assembly, 2 Port, Straight
Catalog # DISI4050-0065
Supplier:  DISTEK
New Product
Vent Assembly, 2 Port, Straight
Catalog # DISI4050-0065
Supplier:  DISTEK
New Product
Specifications
  • Thread:
    GL-45
  • Description:
    Vent assembly, 2-port, straight, universal, straight, GL 45, external hose barb: 6,35 mm
  • Material:
    Stainless steel 316