Order Entry
Northern Ireland
ContactUsLinkComponent
Millipore® Immobilon®-NC Hydrophilic Transfer Membrane
Millipore® Immobilon®-NC Hydrophilic Transfer Membrane
Catalog # MILFHATF85R
Supplier:  Merck
New Product
undefined
Millipore® Immobilon®-NC Hydrophilic Transfer Membrane
Catalog # MILFHATF85R
Supplier:  Merck
New Product

Specifications

  • Description:
    Immobilon®-NC transfer membrane
  • Dimensions:
    70×84 mm

Specifications

About this item

Triton-free, mixed cellulose esters transfer membrane.

  • Western blotting

    Electrostatic

    Hydrophilic

    Mixed cellulose esters (MCE)

    Plain/white filter

    1 roll, 8,5 cm × 10 m

    0,45 µm pore size

Detection method: chromogenic, radioactive and fluorescent.

Compatible stains: Amido black, India ink, Ponceau-S red, Colloidal gold and CPTS.