NANOCOLOR® Photometric Tube Test Kit for Molybdenum
Catalog # MANA985056
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Chemical method:Thioglycolic acid
- Pk:1 KIT
- Description:NANOCOLOR® Molybdenum 40, test no. 0-56
- Dimensions:160×110×119 mm (including packaging)
- Model:MANA985056
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):24
- Weight:327.5 g (including packaging)
- For unit:UV/VIS II; VIS II; 500 D; PF-12Plus; Advance
- Tests per kit:20
- Range:1 - 40,0 mg/L Mo(VI); 1,6 - 65,0 mg/L (MoO₄²⁻)
Specifications
About this item
Photometric test kit for the determination of molybdenum in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Coloured pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: No
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Delivery: Rugged box with 20 test tubes.