ContactUsLinkComponent
Mouse MDC ELISA Kit (Macrophage Derived Chemokine)
Mouse MDC ELISA Kit (Macrophage Derived Chemokine)
Catalog # ABCAAB193726-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse MDC ELISA Kit (Macrophage Derived Chemokine)
Catalog # ABCAAB193726-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse MDC ELISA Kit (Macrophage Derived Chemokine)
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse MDC ELISA Kit (Macrophage Derived Chemokine) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse MDC (Macrophage Derived Chemokine) in Mouse in Plasma, Cell culture supernatant, Serum samples.