Order Entry
Northern Ireland
ContactUsLinkComponent
STAT 5 A/B (pY694/699 + Total) ELISA Kit
STAT 5 A/B (pY694/699 + Total) ELISA Kit
Catalog # ABCAAB205715-96
Supplier:  Abcam
CAS Number:  
undefined
STAT 5 A/B (pY694/699 + Total) ELISA Kit
Catalog # ABCAAB205715-96
Supplier:  Abcam
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    STAT 5 A/B (pY694/699 + Total) ELISA Kit
  • Tests per kit:
    96 tests

Specifications

About this item

STAT 5 A/B (pY694/699 + Total) ELISA Kit is a single-wash 90-min Simplestep used to quantify STAT 5 A/B (pY694/699 + Total) with a sensitivity of 5 µg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.