Order Entry
Northern Ireland
ContactUsLinkComponent
Human Anti-Herpes simplex virus Type 1 IgM ELISA Kit (HSV1)
Human Anti-Herpes simplex virus Type 1 IgM ELISA Kit (HSV1)
Catalog # ABCAAB108738-96
Supplier:  Abcam
CAS Number:  
undefined
Human Anti-Herpes simplex virus Type 1 IgM ELISA Kit (HSV1)
Catalog # ABCAAB108738-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Human Anti-Herpes simplex virus Type 1 IgM ELISA kit (HSV1)
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    96 tests

Specifications

About this item

Human Anti-Herpes simplex virus Type 1 IgM ELISA kit (HSV1) is a Indirect ELISA kit for the measurement of Human Anti-Herpes simplex virus Type 1 IgM (HSV1) in Human in citrate plasma, heparin plasma, serum samples.