ContactUsLinkComponent
Mouse Clusterin ELISA Kit (Apolipoprotein J)
Mouse Clusterin ELISA Kit (Apolipoprotein J)
Catalog # ABCAAB199079-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse Clusterin ELISA Kit (Apolipoprotein J)
Catalog # ABCAAB199079-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse Clusterin ELISA Kit (Apolipoprotein J)
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse Clusterin ELISA Kit (Apolipoprotein J) is a single-wash 90-min Simplestep used to quantify Mouse Clusterin (Apolipoprotein J) with a sensitivity of 280 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.