Mouse EGF ELISA Kit
Catalog # ABCAAB100679-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:EGF
- Description:Mouse EGF ELISA kit
- Sample type:Plasma, cell culture supernatant, serum
- Detection method:Colorimetric
- Detection range:3,28 - 800 pg/ml
- Storage temperature:4 °C
- Sensitivity:3 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Mouse EGF ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse EGF in Mouse in plasma, cell culture supernatant, Serum samples.