ContactUsLinkComponent
Human Complement C3 ELISA Kit
Human Complement C3 ELISA Kit
Catalog # ABCAAB108822-96
Supplier:  Abcam
CAS Number:  
undefined
Human Complement C3 ELISA Kit
Catalog # ABCAAB108822-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human Complement C3 ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Human Complement C3 ELISA kit ab108822 is used to quantify Human Complement C3 with a sensitivity of 8 pg/ml. Competitive ELISA kit.