- Description:Mouse SAP ELISA Kit (PTX2)
- Tests per kit:96 tests
Mouse SAP ELISA Kit (PTX2) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse SAP (PTX2) in Mouse in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum samples.