ContactUsLinkComponent
Human Semaphorid 4D/CD10 ELISA Kit
Human Semaphorid 4D/CD10 ELISA Kit
Catalog # ABCAAB282868-1X96
Supplier:  Abcam
CAS Number:  
undefined
Human Semaphorid 4D/CD10 ELISA Kit
Catalog # ABCAAB282868-1X96
Supplier:  Abcam
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Human Semaphorid 4D/CD10 ELISA kit
Specifications
About this item
Human Semaphorid 4D/CD10 ELISA kit is a single-wash 90-min Simplestep used to quantify Human Semaphorid 4D/CD10 with a sensitivity of 22.9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.