Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Human TNF alpha ELISpot Pair
- Species reactivity:Human
- Sample type:Suspension cells
- Storage temperature:4 °C
- Processing time:Multiple steps
- Antigen symbol:TNF alpha
- Product type:Uncoated
- Tests per kit:10×96 tests
Specifications
About this item
Human TNF alpha ELISpot Pair is a ELISpot Pair for the sensitive detection of Human TNF alpha production in a single cell suspension.