Mouse Complement C3 ELISA Kit
Catalog # ABCAAB157711-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Complement C3 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Complement C3 ELISA kit ab157711 is a sandwich ELISA used to quantify Mouse Complement Factor 3 (C3) with a sensitivity of 1,65 ng/ml. Wide dynamic range – quantifies 3 to 200 ng/ml - Validated in plasma and serum - Colorimetric assay -450 nm readout -works on any plate reader - Cited in over 30 publications.