ContactUsLinkComponent
24(S)-Hydroxycholesterol ELISA Kit
24(S)-Hydroxycholesterol ELISA Kit
Catalog # ABCAAB204530-96
Supplier:  Abcam
CAS Number:  
undefined
24(S)-Hydroxycholesterol ELISA Kit
Catalog # ABCAAB204530-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    24(S)-Hydroxycholesterol ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
24(S)-Hydroxycholesterol ELISA Kit is a Competitive ELISA kit for the measurement of 24(S)-Hydroxycholesterol in Cerebral Spinal Fluid, Tissue Homogenate, Tissue Culture Media samples.