Human DKK1 ELISA Kit (Dickkopf-1)
Catalog # ABCAAB100501-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human DKK1 ELISA kit (Dickkopf-1)
Specifications
About this item
Human DKK1 ELISA kit (Dickkopf-1) is a Sandwich (quantitative) ELISA kit for the measurement of Human DKK1 (Dickkopf-1) in Human in plasma, cell culture supernatant, serum samples.